Cardiac Muscle, Thin Section Slide
Catalog # 470183-666
Supplier: Avantor
Specifications
- Description:Cardiac Muscle, Thin Section Slide
- Environmentally Preferable:
- Cat. No.:470183-666
- Stain:Iron Hematoxylin
- Supplier no.:WARD470183-666
- Orientation:Thin section
Specifications
About this item
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Shows branching fibers, striations, and nuclei.