Order Entry
United States
ContactUsLinkComponent
Carbon Cycle Poster
Carbon Cycle Poster
Catalog # 470004-714
Supplier:  Avantor
undefined
Carbon Cycle Poster
Catalog # 470004-714
Supplier:  Avantor

Specifications

  • Width:
    36"
  • Description:
    Carbon Cycle Poster
  • Height:
    24"
  • Environmentally Preferable:
  • Cat. No.:
    470004-714
  • Supplier no.:
    4730501

Specifications

About this item

A Perfect Lead-in to Lessons on Global Warming

Exceptional full color poster tracks the flow of carbon through the biotic and abiotic parts of the ecosystem. It includes illustrations of respiration, combustion, photosynthesis, mimicry, sedimentation, and decay. Size of poster: 24" x 36".