ContactUsLinkComponent
Areolar Tissue Slide
Areolar Tissue Slide
Catalog # 470177-570
Supplier:  Avantor
undefined
Areolar Tissue Slide
Catalog # 470177-570
Supplier:  Avantor
Specifications
  • Description:
    Areolar Tissue
  • Environmentally Preferable:
  • Cat. No.:
    470177-570
  • Stain:
    Verhoeff and Eosin
  • Supplier no.:
    WARD470177-570
  • Orientation:
    Whole mount
Specifications
About this item
Illustrates specific tissue characteristics
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Spread film of fasciae. Collagenous and elastic fibers.