Specifications
- Color:Black
- Size:4.11" x 200'
- Environmentally Preferable:
- Cat. No.:89007-934
- For use with:GlobalMark Printer
- Supplier no.:96089
Specifications
About this item
Brady has super-tough smear-proof ribbons assure your printed text lasts for years outdoors and in an industrial environment
Ribbons ship self-contained in cartridges that drop straight in to the GlobalMark printer with a "click". This Ribbon is exclusive to use with the Brady RTK labels. Patent D481,7555. RoHS compatibility: compliant with RoHS Directive. NOTE: All statements concerning RoHS Directive compliance refer to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC. Product compliance is based upon information provided by suppliers of the raw materials used by Brady to manufacture these products, or by independent laboratory testing of these products. As such, Brady makes no independent representations or warranties, express or implied, and assumes no liability in connection with the use of this information.