VFlex™ N95 Particulate Respirators, 3M™
Supplier: 3M
Certificates
About this item
Respirators feature unique embossed ‘V’ shapes on the front panel to keep filter media away from the mouth.
- Pleats expand to provide a comfortable seal while giving a spacious feel
- Pleats flex with mouth movement for easier talking
- Tabs on the side are designed as handles
- Compatible with a variety of 3M eyewear and hearing protection
Tabs on the side are designed as handles for easy positioning of the respirator on the face to achieve a comfortable fit. Fold flat for convenient storage. The respirators are NIOSH approved.
Frequently Bought Together