To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Sheep
- Western Blot:Yes
- Size:1 mg
- Environmentally Preferable:
- Epitope:Heavy and Light chain
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Ambient
- Immunogen:Sheep IgG whole molecule
- Tested Applications:Dot Blot
- Purification:purified by immunoaffinity chromatography
- Cat. No.:RL613-4328
- Supplier no.:613-4328
Specifications
About this item
Suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays.
Recommended Dilutions: ELISA: 1:5000 - 1:22000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:300 - 1:1500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Sheep