To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Pack type:Fiber drum
- CAS No.:8049-47-6
- Source:
- Species:
- Size:50 kg
- Environmentally Preferable:
- Storage Conditions:Refrigerator
- Form:Powder
- Enzyme Name:Pancreatin
- Grade:USP
- Cat. No.:700011-472
- Supplier no.:PA115-50KGBL
Form: Powder
Pancreatin, powder, USP is used to replace missing pancreatic enzymes and reduces iron absorption from food in the duodenum during the digestion process. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.