VWR® Protective Eyewear with Adjustable Temples
Catalog # 89187-984
Supplier: VWR International
CSA Certified
Product available on GSA Advantage®
Specifications
- Frame color:Black
- Environmentally Preferable:
- Cat. No.:89187-984
- Lens coating:Anti-fog, scratch-resistant
- Lens tint:Clear
Specifications
About this item
Ultra-lightweight nylon frame with soft PVC nose pads.
- Unique ratcheting/adjustable temples lock into place for a custom fit
- Ultra-lightweight nylon frames and soft PVC nose pads to prevent slipping
- 8-base spherical lenses provide excellent side protection
- Anti-fog coating extends lens life
Wrap-around sport styling, designed for men and women features eight base spherical lenses providing excellent side protection. The impact resistant polycarbonate lenses provide 99.9% UV protection and the scratch resistant coating.
Certifications: ANSI Z87.1