To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Thyroid Hormone Receptor, Beta
- Antigen Symbol:THRB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:25 µL
- Environmentally Preferable:
- Epitope:N-Terminal
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:7068
- Antigen Synonyms:GRTH|ERBA-BETA|THRB2|NR1A2|ERBA2|MGC126109|thyroid hormone receptor|avian)|oncogene ERBA2|THRB1|beta|MGC126110|THR1|thyroid hormone receptor beta 1|avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2|generalized resistance to thyroid hormone|beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2|Thyroid hormone receptor
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.66 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:This TR Beta1 antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the N-terminal of human THRB isoform 1 Protein.
- Purification:affinity-purified
- Cat. No.:RL600-401-A96S
- Supplier no.:600-401-A96S
Specifications
About this item
This affinity purified antibody has been tested for use in ELISA and western blotting.
Recommended Dilutions: ELISA: 1:200,000 – 1:220000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: THRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human