To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Tag
- Antigen Name:Green Fluorescence Protein
- Antigen Symbol:GFP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western Blot:Yes
- Size:25 µL
- Environmentally Preferable:
- Epitope:Full length
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen Synonyms:GFPuv|eGFP|Green Fluorescent Protein
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:0.66 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:The immunogen is a Green Fluorescent Protein (GFP) fusion Protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria.
- Tested Applications:Fluoresence ELISA
- Purification:purified by immunoaffinity chromatography
- Cat. No.:RL600-901-215S
- Supplier no.:600-901-215S
Specifications
About this item
Anti-GFP is designed to detect GFP and its variants. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen and recognizes wild type, recombinant and enhanced forms of GFP.
Recommended Dilutions: ELISA: 1:2000 - 1:12000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:200 - 1:1000;
Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Chicken
Isotype: IgG
Reactivity: