Halotron® I Fire Extinguishers, Kidde
Supplier: Avantor
Certificates
About this item
Upright fire extinguisher for B and C fires. The extinguishing material Halotron® is an acceptable alternative to Halon I2II.
- Leaves no residue
- Agent will not cause thermal shock
- Class A capability
- High performance clean agent
- Steel housing, metal valve, handle and lever
Ordering information: Wall hanger included.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.