Bosch Power Tools Self-Leveling Cross-Line Laser Level, ORS Nasco
300076-503
: Avantor
- Description:Self-Leveling Cross-Line Laser Level
- Environmentally Preferable:
- Cat. No.:300076-503
Smart Pendulum Leveling System self-levels, senses, and indicates out-of level conditions
- Vertical, horizontal, and crossLine modes
- 65' range
- Requires (3) AA batteries
- 635nm laser wavelength
- ±0.25" accuracy
Will also lock when switched off. Projects two lines independently or together for a wide array of leveling and/or plumb applications.
: Device includes batteries, belt pouch, carrying case, and WM1 positioning device.