Order Entry
United States
ContactUsLinkComponent
Compact Cable Cutter, Klein Tools
Compact Cable Cutter, Klein Tools
  300021-154
 :  Avantor
undefined
Compact Cable Cutter, Klein Tools
  300021-154
 :  Avantor

 

  • Environmentally Preferable:
  • Cat. No.:
    300021-154
  • Cutting capacity:
    24 gauge
  • Overall length:
    8"
  • Weight:
    0.73 lbs.
  • Supplier no.:
    409-63055
  • Grip type:
    Plastic-dipped
  • Cut type:
    Shear cut, straight

 

 

Cable cutter with heat-treated, high-carbon steel blades. Mainly used for cutting multi-strand cable and wire, such as 24-gauge, 100 pair telephone cable.

  • Drop forged body with 3/8" case hardened rivet
  • One hand shearing action

Shaer angle cut offers clean cut without compression or fraying ends.