Order Entry
United States
ContactUsLinkComponent
Aqua-Poly/Mount, Polysciences
Aqua-Poly/Mount, Polysciences
Catalog # 87001-902
Supplier:  Polysciences Inc
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:  
undefined
Aqua-Poly/Mount, Polysciences
Catalog # 87001-902
Supplier:  Polysciences Inc
Supplier Number:  18606-20
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Aqua-Poly/Mount
  • Size:
    20 mL (0.68 oz.)
  • Environmentally Preferable:
  • Cat. No.:
    87001-902
  • Supplier no.:
    18606-20

Specifications

About this item

Water soluble, non-fluorescing mounting medium formulated for mounting sections from aqueous solutions, frozen sections, and special procedures that will not tolerate alcohols or solvents

Ideal for immunofluorescent techniques as it enhances and retains fluorescent stains. Coverslip directly from water or buffer using standard methods