L(+)-Glutamic acid free acid, high purity
Catalog # 97061-634
Supplier: VWR
CAS Number:
Specifications
- Size:1 kg
- Environmentally Preferable:
- Cat. No.:97061-634
- Supplier no.:0421-1KG
Specifications
Specification Test Results
- Heavy Metals (as Pb):= 0.002 %
- Lead:= 0.001 %
- Loss on Drying:= 0.1 %
- Purity (Dried):98.5 % - 101.5 %
- Specific Rotation (Dried):31.5 ° - 32.5 °
Specification Test Results
About this item
Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
:
:147.13 g/mol
CAS Number:56-86-0
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number:MFCD00002634
Boiling Pt:325
Melting Pt:160
Merck Index:13,04482