Methanol absolute ≥99.8% ACS for synthesis
Supplier: Spectrum Chemicals
Certificates
About this item
Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
Boiling Pt:64.6
Melting Pt:-98
Density:0.787
Molecular Weight:32.04 g/mol
Flash Pt:11 °C (closed cup)
UN:1230
ADR:3 II
Merck Index:13,05984
Storage Temperature:Ambient
EINECS:200-659-6
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.8% Min.
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_M1240
SDS
M1240 spec
Product specification
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together