D-(+)-Glucose 40% (w/v) in aqueous solution, Ultra Pure Grade for biochemistry
Catalog # 97062-946
Supplier: VWR
CAS Number:
Specifications
- Pack type:Bottle, clear
- Size:100 ml
- Environmentally Preferable:
- Cat. No.:97062-946
- Supplier no.:E701-100ML
Specifications
Specification Test Results
- Appearance:Clear to slightly yellow liquid
- Glucose:35 % - 40 %
Specification Test Results
About this item
Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
:
:180.16 g/mol
EINECS:207-757-8
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
Melting Pt:146
Merck Index:14,04459