Order Entry
United States
ContactUsLinkComponent
 
Calcium acetate hemihydrate ≥99%, high purity
Catalog # 97061-232
Supplier:  Avantor
CAS Number:  
undefined
Calcium acetate hemihydrate ≥99%, high purity
Catalog # 97061-232
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    HDPE bottle
  • Size:
    500 g
  • Environmentally Preferable:
  • Cat. No.:
    97061-232
  • Supplier no.:
    0225-500G

Specifications

Specification Test Results

  • Chloride:
    0.01 %
  • Heavy Metals:
    0.0025 %
  • Loss on Drying:
    = 7 %
  • Purity:
    = 99 %

Specification Test Results

About this item

Synonyms: Calcium di(acetate), Acetic acid calcium salt
 :
 :167.18 g/mol
Melting Pt:160 °C
Density:1.509 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-540-9
CAS Number:62-54-4
Formula:(H₃CCOO)₂Ca·¹/₂H₂O