- Description:Evac Waste System Supplied with 4L Polypropylene Bottle and Cap with Level Detection and Quick Release Couplings
- Environmentally Preferable:
- Electrical:110–240V
- Cat. No.:89129-550
- Supplier no.:EV310
These compact waste systems are ideal for use with critical or dangerous liquids or supernatants, pathogens, or any class III or IV biohazard laboratories
E-Vac is available with a 4 L polypropylene bottle or a 3 L safety glass bottle. Polypropylene and glass bottles offer lids equipped with quick-release tubing connectors for safe and convenient tubing connections; and level detection sensor that automatically shuts the pump off when the bottle is full. Polypropylene bottle also available with lid fitted with standard barbed tube fittings.
EV310 includes 4 L polypropylene bottle, lid with quick release couplings and level detection sensor, tubing set with filter, connector adapter 100-240V, 50/60Hz, HandE-Vac Hand Operator, includes single channel adapter for disposable tips.
EV320 includes 4 L polypropylene bottle, lid with barbed tube fittings, tubing set with filter, connector adapter 100-240V, 50/60Hz, HandE-Vac Hand Operator, includes single channel adapter for disposable tips.
Quiet, oil-free membrane pump has a range from -250mm to 650 mm Hg. Once target vacuum is reached, pump automatically switches off. Pump automatically switches on when vacuum pressure is applied allowing it to maintain a constant vacuum pressure – perfect for gentle aspiration of small plates and dishes, as well as rapid emptying of larger containers. Easy-to-clean stainless steel housing is UV-resistant for use in laminar hoods.