About this item
Clarity Urine Controls for use with Siemens/Roche Urine Strips and Siemens/Roche Urine analyzers only.
- Dropper style controls
- Color coded cap: Blue cap for negative and red cap for positive
- No refrigeration required in transit
- Equivalent to BioRad and Quantimetrix urine controls
For use with Siemens and Roche Micro albumin creatine test strips. For use with all Rapid hCG/Pregnancy tests including Clarity hCG Tests.
Comes in easy-dispensing dropper bottles, and remains stable in a liquid state for two years, even after being opened.