Order Entry
United States
ContactUsLinkComponent
APC® Power-Saving Essential SurgeArrest Surge Protector, Essendant
APC® Power-Saving Essential SurgeArrest Surge Protector, Essendant
  500040-609
 :  Avantor
undefined
APC® Power-Saving Essential SurgeArrest Surge Protector, Essendant
  500040-609
 :  Avantor

 

  • Response time:
    1 ns
  • Cord length:
    4 '
  • No. of outlets:
    7
  • Depth:
    1.40 "
  • Width:
    2.80 "
  • Housing material:
    Plastic
  • Joules of surge protection:
    1020 J