Navigator® Premium Recycled Multipurpose Paper
500018-751
: Avantor
- Color:White
- Dimensions:8.5×11"
- Environmentally Preferable:
- Cat. No.:500018-751
- Weight:20 lbs.
- Supplier no.:SNANR1120
Quality high bright white paper for great impressions.
- Brightness rating of 110 (International) and 95 (US)
Ideal for everyday printing. Specially engineered to deliver high-quality results. Suitable for multifunction, laser and inkjet printers, fax machines, copiers and offset printing presses.