Canon® Ink Tank, CLI36, Essendant LLC MS
Catalog # 500014-536
Supplier: Janitorial Supplies
Specifications
- Cartridge no.:CLI-36
- For Use With:Inkjet Printer
- Yield:100 pages
- Color:Tri-Color
- Environmentally Preferable:
- Cat. No.:500014-536
- Supplier no.:CNMCLI36
Specifications
About this item
Maximizes the potential of your Canon® printer
- OEM and not remanufactured
Creates sharp lines and crisp pages. Every cartridge is simple to install and use.