Order Entry
United States
ContactUsLinkComponent
 
HP Bright White Inkjet Paper
  500019-682
undefined
HP Bright White Inkjet Paper
  500019-682
 :  HEW203000

 

  • For Use With:
    Inkjet Printers
  • Color:
    Bright white
  • Description:
    Super bright paper, brightness rating: Int. 108/US 98
  • Dimensions:
    8.5×11"
  • Environmentally Preferable:
  • Cat. No.:
    500019-682
  • Packaging:
    500 sheets/unit
  • Weight:
    24 lbs.
  • Supplier no.:
    HEW203000

 

 

An extra bright paper formulated to enhance the output of color inkjet printers

Special surface enhancements fix inks onto the sheet. ColorLok™ technology for smear-resistance, bolder blacks and brighter colors.

 : FSC Certified.