Order Entry
United States
ContactUsLinkComponent
HP Premium Choice Laser Paper
HP Premium Choice Laser Paper
Catalog # 500019-675
undefined
HP Premium Choice Laser Paper
Catalog # 500019-675
Supplier Number:  HEW113100

Specifications

  • For Use With:
    HP Color LaserJet Printers & Color Copiers, Copiers, Laser Printers
  • Color:
    Ultra white
  • Dimensions:
    8.5×11"
  • Environmentally Preferable:
  • Cat. No.:
    500019-675
  • Packaging:
    500 sheets/unit
  • Weight:
    32 lbs.
  • Finish:
    Super smooth
  • Supplier no.:
    HEW113100

Specifications

About this item

The one paper to run your office smarter! Because you can use it in any office machine, there's no need to keep specialty papers on hand for everyday use.

  • Brightness rating 112 (International) and 98 (US)
  • Acid-free for archival quality

It's brighter than ordinary copy paper, with an extra smooth surface that produces sharp, high-contrast images.