Texas Instruments TI-1706SV Handheld Pocket Calculator
500003-873
- Description:Solar pocket calculator
- Dimensions:3×5"
- Environmentally Preferable:
- Cat. No.:500003-873
- Supplier no.:TEXTI1706SV
Well equipped with SuperView display and solar/battery power for use anywhere
- Wall mountable
- Color-coded keyboard with extra-large, easy-to-use keys
- LCD displays clear 8 digits (11.5 mm height) for easy readability
- Change sign (+/–) key simplifies entry of negative numbers
Protective slide case stores on back until needed. Features percent key, change sign (+/–) key, square root key, percent add-on/discount and 3-key memory.