DAX® Cubicle Photo/Document Frame
Catalog # 500012-636
Supplier: Avantor
Specifications
- Color:Clear
- Dimensions:8¹/₂W×11"H
- Material:Acrylic
- Environmentally Preferable:
- Cat. No.:500012-636
- Supplier no.:DAXN140285M
Specifications
About this item
VELCRO® piece on back sticks to fabric wall, or use the VELCRO® backed magnet that is included.
- Clear acrylic frame displays and protects photos/documents
- Contains 70% recycled material
Hang frame on file cabinets or other metal surfaces either vertically or horizontally. Velcro piece on back sticks to fabric wall.
This frame is used for document/certificates, and also it is a wall mounting frame.