ContactUsLinkComponent
Sharpie® Industrial Permanent Marker
Sharpie® Industrial Permanent Marker
Catalog # 500034-938
undefined
Sharpie® Industrial Permanent Marker
Catalog # 500034-938
Supplier Number:  SAN13601
Specifications
  • Color:
    Black
  • Environmentally Preferable:
  • Cat. No.:
    500034-938
  • Supplier no.:
    SAN13601
Specifications
About this item
For industrial, laboratory and commercial use.
  • With fine tip and non-washable ink
Tough under extreme conditions-even at 500 °C. Great for metal, foil, glass, film and plastic. Fade and water-resistant black ink.