AquaSep™ Premier Base Deactivated Phase HPLC Columns, 100 Å, ES Industries
Supplier: Avantor
Certificates
About this item
AquaSep is the clear choice for the discriminating HPLC chromatographer faced with a difficult separation challenge such as polar compounds, compounds requiring a highly aqueous mobile phase or difficult to retain compounds.
The AquaSep phase has been specially developed using patented technology for use with highly aqueous mobile phases, including 100% aqueous. Our unique patented approach provides a complete solution to ensure that AquaSep is totally resistant to 'phase collapse' under all mobile phase conditions.