ContactUsLinkComponent
Barnstead B-pure™ Cartridge Water Purification System, Thermo Scientific
undefined
Barnstead B-pure™ Cartridge Water Purification System, Thermo Scientific
  • Catalog No:
  • 26302-250
  • 82002-642
  • 26302-448
  • 26302-253
  • Max. Flow Rate:
  • 4 L per minute
  • 4 L per minute
  • 4 L per minute
  • 4 L per minute
  • Max. Inlet Pressure:
  • 100 psig
  • 100 psig
  • 100 psig
  • 100 psig
  • Feed Water:
  • 4 to 49°C (40 to 120°F)
  • 4 to 49°C (40 to 120°F)
  • 4 to 49°C (40 to 120°F)
  • 4 to 49°C (40 to 120°F)
  • Inlet/Outlet Connection:
  • 12.7 mm (1/2") NPTF
  • 12.7 mm (1/2") NPTF
  • 12.7 mm (1/2") NPTF
  • 12.7 mm (1/2") NPTF
Frequently Bought Together