Brownlee™ Aquapore® MPLC™ Columns (300 Å Pore Size), PerkinElmer
Supplier: Avantor
Certificates
About this item
These columns are packed with a wide-pore silica-based sorbent that is suitable for the separation of large biopolymers.
- Wide pore design
- Silica-based sorbent
- Suitable for the separation of large biopolymers
Columns are available in an extensive range of dimensions and particle sizes for use in micro-scale separations, LC/MS analyses, and high-speed preparative analyses up to process scale.
Frequently Bought Together