Lindberg/Blue M Box Furnaces, 1200 °C, Thermo Scientific
Supplier: Avantor
Certificates
About this item
Box furnaces provide uniform heat distribution through individual heating elements on all sides of chamber.
- Door conveniently opens to horizontal position to serve as chamber loading surface
- Spring-loaded mechanism holds door securely shut
- A sight glass area allows convenient observation of the heated sample when furnace is in operation
- Power to furnace is shut off if high limit is reached
An insulated vestibule and tight-fitting insulated door plug provide excellent temperature uniformity by minimizing heat loss around the furnace chamber opening. Over-temperature protection feature on controller 30783-004 protects furnace and sample if primary control circuit fails. Maximum temperature is 1200°C. For 208/240V, 50/60Hz.
Ordering information: Furnaces feature a one-year parts and labor warranty. Includes Platinel® II thermocouple with 10' compensated lead wire and polarized plug. Controller required for operation, sold separately. Power cord and interconnecting wiring not included.
Frequently Bought Together