ContactUsLinkComponent
Whatman™ PolyVENT™ Integral Vent Filters
Whatman™ PolyVENT™ Integral Vent Filters
Supplier:  Cytiva
Certificates
undefined
Whatman™ PolyVENT™ Integral Vent Filters
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • 28137-652
  • 28137-653
  • 28137-658
  • 28137-650
  • 28137-656
  • Max. Pressure:
  • 2 bar - forward direction (29 psi - forward direction)
  • 2 bar - forward direction (29 psi - forward direction)
  • 2 bar - forward direction (29 psi - forward direction)
  • 2 bar - forward direction (29 psi - forward direction)
  • 2 bar - forward direction (29 psi - forward direction)
  • Application:
  • Venting (filling).
    Isolation (incubators, autoclaves, lyophilizers, EtO sterilizers, fermenters).
    Electronics (gases).
  • Venting (filling).
    Isolation (incubators, autoclaves, lyophilizers, EtO sterilizers, fermenters).
    Electronics (gases).
  • Venting (filling).
    Isolation (incubators, autoclaves, lyophilizers, EtO sterilizers, fermenters).
    Electronics (gases).
  • Venting (filling).
    Isolation (incubators, autoclaves, lyophilizers, EtO sterilizers, fermenters).
    Electronics (gases).
  • Venting (filling).
    Isolation (incubators, autoclaves, lyophilizers, EtO sterilizers, fermenters).
    Electronics (gases).
  • Filter Material:
  • Polytetrafluoroethylene (PTFE)
  • Polytetrafluoroethylene (PTFE)
  • Polytetrafluoroethylene (PTFE)
  • Polytetrafluoroethylene (PTFE)
  • Polytetrafluoroethylene (PTFE)