ContactUsLinkComponent
Acro® 50 Vent Filters with Emflon® II Membrane, Cytiva (Formerly Pall Lab)
Acro® 50 Vent Filters with Emflon® II Membrane, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
Certificates
undefined
Acro® 50 Vent Filters with Emflon® II Membrane, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • 28143-838
  • 16003-656
  • Filtration Area:
  • 20 cm²
  • 20 cm²
  • Max. Ambient Temperature:
  • 60°C (140°F)
  • 60°C (140°F)
  • Max. Operating Pressure:
  • 4.1 bar (60 psi) at ambient temperature
  • 4.1 bar (60 psi) at ambient temperature