ContactUsLinkComponent
Target® Syringe Filters, PVDF, Thermo Scientific
Target® Syringe Filters, PVDF, Thermo Scientific
Supplier:  Thermo Scientific
Certificates
undefined
Target® Syringe Filters, PVDF, Thermo Scientific
Supplier:  Thermo Scientific
Specifications
  • Catalog No:
  • 66064-433
  • 66064-832
  • 66030-847
  • 66064-397
  • 66030-867
  • 66030-880
  • Max. Operating Pressure:
  • 75 psi for 4 mm, 90 psi for 30 mm
  • 75 psi for 4 mm, 90 psi for 30 mm
  • 75 psi for 4 mm, 90 psi for 30 mm
  • 75 psi for 4 mm, 90 psi for 30 mm
  • 75 psi for 4 mm, 90 psi for 30 mm
  • 75 psi for 4 mm, 90 psi for 30 mm
  • Max. Temperature:
  • 110°C (230°F)
  • 110°C (230°F)
  • 110°C (230°F)
  • 110°C (230°F)
  • 110°C (230°F)
  • 110°C (230°F)