Anti-AP2A1 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:20-1:100
Type:
Antigen: AP2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77705-879
- 77705-880
- Antigen Symbol:
- AP2A1
- AP2A1
- Antigen Synonyms:
- AP-2 complex subunit alpha-1|adaptor protein complex AP2 subunit alpha 1|Clathrin assembly protein complex 2 alpha A large chain|adapter-related protein complex 2 subunit alpha-1|CLAPA1|Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit|Alpha adaptin A|Adaptin alpha|AP2-ALPHA|alpha1 adaptin|Clathrin assembly protein complex 2 alpha-A large chain|clathrin associated/assembly/adaptor protein, large, alpha 1|Adaptin alpha A|Adaptor protein complex AP-2 subunit alpha-1|Adaptor protein complex AP2 alpha 1 subunit|Adaptor protein complex AP 2 alpha 1 subunit|CLAPA 1|clathrin associated assembly adaptor protein large alpha 1|Adaptor related protein complex AP2 alpha 1 subunit|Alpha1-adaptin|ADTAA|Alpha-adaptin A|Adaptor related protein complex AP 2 alpha 1 subunit|Clathrin associated / assembly / adaptor protein large alpha 1|Plasma membrane adaptor HA 2/AP 2 adaptin alpha A subunit|Alpha A adaptin|AP 2 complex subunit alpha 1|Adapter-related protein complex 2 alpha-1 subunit|adaptor protein complex AP 2 subunit alpha 1|AP 2 alpha|AP2 complex subunit alpha 1|Adaptor related protein complex 2 alpha 1 subunit|AP2A1_HUMAN|100 kDa coated vesicle protein A|Clathrin adaptor complex AP2. alpha subunit|Ap2a1
- AP-2 complex subunit alpha-1|adaptor protein complex AP2 subunit alpha 1|Clathrin assembly protein complex 2 alpha A large chain|adapter-related protein complex 2 subunit alpha-1|CLAPA1|Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit|Alpha adaptin A|Adaptin alpha|AP2-ALPHA|alpha1 adaptin|Clathrin assembly protein complex 2 alpha-A large chain|clathrin associated/assembly/adaptor protein, large, alpha 1|Adaptin alpha A|Adaptor protein complex AP-2 subunit alpha-1|Adaptor protein complex AP2 alpha 1 subunit|Adaptor protein complex AP 2 alpha 1 subunit|CLAPA 1|clathrin associated assembly adaptor protein large alpha 1|Adaptor related protein complex AP2 alpha 1 subunit|Alpha1-adaptin|ADTAA|Alpha-adaptin A|Adaptor related protein complex AP 2 alpha 1 subunit|Clathrin associated / assembly / adaptor protein large alpha 1|Plasma membrane adaptor HA 2/AP 2 adaptin alpha A subunit|Alpha A adaptin|AP 2 complex subunit alpha 1|Adapter-related protein complex 2 alpha-1 subunit|adaptor protein complex AP 2 subunit alpha 1|AP 2 alpha|AP2 complex subunit alpha 1|Adaptor related protein complex 2 alpha 1 subunit|AP2A1_HUMAN|100 kDa coated vesicle protein A|Clathrin adaptor complex AP2. alpha subunit|Ap2a1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95782
- O95782
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human AP2A1
- Synthetic peptide of Human AP2A1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together