Order Entry
United States
ContactUsLinkComponent
Anti-SMARCD1 Rabbit Polyclonal Antibody (FITC)
Anti-SMARCD1 Rabbit Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
Anti-SMARCD1 Rabbit Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77723-834
  • 77723-833
  • Antigen Symbol:
  • SMARCD1
  • SMARCD1
  • Antigen Synonyms:
  • SMRD1|BRG1 associated factor 60A|BRG1-associated factor 60A|SMRD1_HUMAN|SMARCD1|SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1|60 kDa BRG1/Brm associated factor subunit A|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 1|SWI/SNF complex 60 kDa subunit A|Swp73 like protein|BAF60A|CRACD1|Rsc6p|60 kDa BRG-1/Brm-associated factor subunit A|SWI/SNF related matrix associated actin dependent regulator of chromatin d1|Chromatin remodeling complex BAF60A subunit|SWI/SNF complex 60 kDa subunit|Mammalian chromatin remodeling complex BRG1 associated factor 60A
  • SMRD1|BRG1 associated factor 60A|BRG1-associated factor 60A|SMRD1_HUMAN|SMARCD1|SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1|60 kDa BRG1/Brm associated factor subunit A|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 1|SWI/SNF complex 60 kDa subunit A|Swp73 like protein|BAF60A|CRACD1|Rsc6p|60 kDa BRG-1/Brm-associated factor subunit A|SWI/SNF related matrix associated actin dependent regulator of chromatin d1|Chromatin remodeling complex BAF60A subunit|SWI/SNF complex 60 kDa subunit|Mammalian chromatin remodeling complex BRG1 associated factor 60A
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q96GM5
  • Q96GM5
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 1 protein (7-124AA)
  • Recombinant Human SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 1 protein (7-124AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue Ice
  • Shipped on Blue Ice

Specifications

Frequently Bought Together