Order Entry
United States
ContactUsLinkComponent
Anti-ATP6V0A1 Rabbit Polyclonal Antibody (Biotin)
Anti-ATP6V0A1 Rabbit Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product
Certificates
Anti-ATP6V0A1 Rabbit Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77723-643
  • 77723-644
  • Antigen Symbol:
  • ATP6V0A1
  • ATP6V0A1
  • Antigen Synonyms:
  • V-type proton ATPase 116 kDa subunit a isoform 1|Vacuolar proton pump subunit 1|V ATPase 116 kDa|V ATPase 116 kDa isoform a1|H(+) transporting two sector ATPase 116 kDa accessory protein A1|ATPase H+ transporting lysosomal V0 subunit a1|ATPase H+ transporting lysosomal non catalytic accessory protein 1 (110/116kD)|V type proton ATPase 116 kDa subunit a|Clathrin coated vesicle/synaptic vesicle proton pump 116 kDa subunit|VPP1|Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit|ATPase H+ transporting lysosomal (vacuolar proton pump) non catalytic accessory protein 1A (110/116kD)|V-ATPase 116 kDa isoform a1|ATP6V0A1|a1|Vacuolar type H(+) ATPase 115 kDa subunit|Stv1|V type proton ATPase 116 kDa subunit a isoform 1|Vacuolar proton pump, subunit 1|ATP6N1A|VPP1_HUMAN|Vacuolar proton translocating ATPase 116 kDa subunit A|Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1|Vph1|ATPase H+ transporting V0 subunit a1|DKFZp781J1951|Vacuolar adenosine triphosphatase subunit Ac116|ATP6N1|ATPase, H+ transporting, lysosomal, noncatalytic accessory protein 1A
  • V-type proton ATPase 116 kDa subunit a isoform 1|Vacuolar proton pump subunit 1|V ATPase 116 kDa|V ATPase 116 kDa isoform a1|H(+) transporting two sector ATPase 116 kDa accessory protein A1|ATPase H+ transporting lysosomal V0 subunit a1|ATPase H+ transporting lysosomal non catalytic accessory protein 1 (110/116kD)|V type proton ATPase 116 kDa subunit a|Clathrin coated vesicle/synaptic vesicle proton pump 116 kDa subunit|VPP1|Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit|ATPase H+ transporting lysosomal (vacuolar proton pump) non catalytic accessory protein 1A (110/116kD)|V-ATPase 116 kDa isoform a1|ATP6V0A1|a1|Vacuolar type H(+) ATPase 115 kDa subunit|Stv1|V type proton ATPase 116 kDa subunit a isoform 1|Vacuolar proton pump, subunit 1|ATP6N1A|VPP1_HUMAN|Vacuolar proton translocating ATPase 116 kDa subunit A|Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1|Vph1|ATPase H+ transporting V0 subunit a1|DKFZp781J1951|Vacuolar adenosine triphosphatase subunit Ac116|ATP6N1|ATPase, H+ transporting, lysosomal, noncatalytic accessory protein 1A
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q93050
  • Q93050
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human V-type proton ATPase 116 kDa subunit a isoform 1 protein (80-260AA)
  • Recombinant Human V-type proton ATPase 116 kDa subunit a isoform 1 protein (80-260AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue Ice
  • Shipped on Blue Ice

Specifications

Frequently Bought Together