Anti-PRDX4 Rabbit Polyclonal Antibody
77689-621
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Symbol:PRDX4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q13162
- Antigen Synonyms:Thioredoxin peroxidase AO372|Peroxiredoxin-4|PRX 4|Peroxiredoxin4|PRDX4_HUMAN|PrxIV|PRDX 4|Thioredoxin peroxidase|Thioredoxin-dependent peroxide reductase A0372|PRX4|Antioxidant enzyme AOE372|Peroxiredoxin IV|Antioxidant enzyme 372|Thioredoxin dependent peroxide reductase A0372|AOE372|Prx-IV|EC 1.11.1.15|Prdx4|Thioredoxin Peroxidase (Antioxidant Enzyme)|AOE37 2|TRANK|AOE37-2|Prx IV
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Peroxiredoxin-4 protein (38-271AA)
- Purification:>95%, Protein G purified
- Cat. No.:77689-621
- Packaging:Vial
Recommended dilution: WB:1:500-1:2000
Type:
Antigen: PRDX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human