Order Entry
United States
ContactUsLinkComponent
Anti-PLA2G6 Rabbit Polyclonal Antibody
Anti-PLA2G6 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
Anti-PLA2G6 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77713-059
  • 77713-060
  • Antigen Symbol:
  • PLA2G6
  • PLA2G6
  • Antigen Synonyms:
  • Phospholipase A2 group VI cytosolic calcium independent|iPLA(2)beta|iPLA2-beta|Phospholipase A2, group VI (cytosolic, calcium independent)|PARK14|BB112799|Neurodegeneration with brain iron accumulation 2|Cytosolic calcium independent phospholipase A2|CaI-PLA2|Group VI phospholipase A2|Intracellular membrane associated calcium independent phospholipase A2 beta|PLPL9_HUMAN|Patatin-like phospholipase domain-containing protein 9|INAD1|PNPLA9|iPLA2 beta|PLA2G6|Calcium independent phospholipase A2|85 kDa calcium independent phospholipase A2|Phospholipase A2 calcium independent|NBIA2|Group VI, A|NBIA2A|CTA-228A.2|Intracellular membrane-associated calcium-independent phospholipase A2 beta|Phospholipase A2 group VI|OTTHUMP00000199457|EC 3.1.1.4|Phospholipase A2 calcium independent group VI A|iPLA2beta|CaI PLA2|Patatin like phospholipase domain containing protein 9|NBIA2B|PLA2|IPLA2-VIA|CaIPLA2|85/88 kDa calcium-independent phospholipase A2|Patatin like phospholipase domain containing 9|GVI PLA2|GVI
  • Phospholipase A2 group VI cytosolic calcium independent|iPLA(2)beta|iPLA2-beta|Phospholipase A2, group VI (cytosolic, calcium independent)|PARK14|BB112799|Neurodegeneration with brain iron accumulation 2|Cytosolic calcium independent phospholipase A2|CaI-PLA2|Group VI phospholipase A2|Intracellular membrane associated calcium independent phospholipase A2 beta|PLPL9_HUMAN|Patatin-like phospholipase domain-containing protein 9|INAD1|PNPLA9|iPLA2 beta|PLA2G6|Calcium independent phospholipase A2|85 kDa calcium independent phospholipase A2|Phospholipase A2 calcium independent|NBIA2|Group VI, A|NBIA2A|CTA-228A.2|Intracellular membrane-associated calcium-independent phospholipase A2 beta|Phospholipase A2 group VI|OTTHUMP00000199457|EC 3.1.1.4|Phospholipase A2 calcium independent group VI A|iPLA2beta|CaI PLA2|Patatin like phospholipase domain containing protein 9|NBIA2B|PLA2|IPLA2-VIA|CaIPLA2|85/88 kDa calcium-independent phospholipase A2|Patatin like phospholipase domain containing 9|GVI PLA2|GVI
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O60733
  • O60733
  • Isotype:
  • IgG
  • IgG
  • Immunogen:
  • Synthetic peptide of Human PLA2G6
  • Synthetic peptide of Human PLA2G6
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together