Order Entry
United States
ContactUsLinkComponent
Anti-IL12A Rabbit Polyclonal Antibody (Biotin)
Anti-IL12A Rabbit Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product
Certificates
Anti-IL12A Rabbit Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77711-353
  • 77711-352
  • Antigen Symbol:
  • IL12A
  • IL12A
  • Antigen Synonyms:
  • NKSF|IL 12A|NK cell stimulatory factor chain 2|Interleukin 12 subunit beta|IL12|Interleukin 12B|NK cell stimulatory factor chain 1|Interleukin 12 beta chain|CLMF|Interleukin-12 subunit alpha|IL35 subunit|CLMF2|Interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40)|Cytotoxic lymphocyte maturation factor 1, p35|Cytotoxic lymphocyte maturation factor 35 kDa subunit|NKSF2|CLMF p40|IL-12A|IL 2 subunit p40|IL-12 subunit p35|IL12, subunit p40|IL-12B|Cytotoxic lymphocyte maturation factor 40 kDa subunit|P35|CLMF1|Interleukin 12, p40|Interleukin 12 alpha chain|IL12A|NFSK|IL12A_HUMAN|Natural killer cell stimulatory factor 1, 35 kD subunit|CLMF p35|Interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35)|NKSF1|natural killer cell stimulatory factor 2|Natural killer cell stimulatory factor, 40 kD subunit|IL12, p35 subunit|IL12B|NF cell stimulatory factor chain 1|Cytotoxic lymphocyte maturation factor 2
  • NKSF|IL 12A|NK cell stimulatory factor chain 2|Interleukin 12 subunit beta|IL12|Interleukin 12B|NK cell stimulatory factor chain 1|Interleukin 12 beta chain|CLMF|Interleukin-12 subunit alpha|IL35 subunit|CLMF2|Interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40)|Cytotoxic lymphocyte maturation factor 1, p35|Cytotoxic lymphocyte maturation factor 35 kDa subunit|NKSF2|CLMF p40|IL-12A|IL 2 subunit p40|IL-12 subunit p35|IL12, subunit p40|IL-12B|Cytotoxic lymphocyte maturation factor 40 kDa subunit|P35|CLMF1|Interleukin 12, p40|Interleukin 12 alpha chain|IL12A|NFSK|IL12A_HUMAN|Natural killer cell stimulatory factor 1, 35 kD subunit|CLMF p35|Interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35)|NKSF1|natural killer cell stimulatory factor 2|Natural killer cell stimulatory factor, 40 kD subunit|IL12, p35 subunit|IL12B|NF cell stimulatory factor chain 1|Cytotoxic lymphocyte maturation factor 2
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P29459
  • P29459
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Interleukin-12 subunit alpha protein (23-219AA)
  • Recombinant Human Interleukin-12 subunit alpha protein (23-219AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together