Anti-COL4A3BP Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:100-1:300
Type:
Antigen: COL4A3BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77716-021
- 77716-022
- Antigen Symbol:
- COL4A3BP
- COL4A3BP
- Antigen Synonyms:
- Cert protein|FLJ20597|C43BP_HUMAN|Ceramide transporter|col4a3bp|hCERT|Alpha 3 type IV collagen binding protein|StARD 11|Procollagen type IV alpha 3 (Goodpasture antigen) binding protein|CERT|Collagen type IV alpha 3 binding protein|CERTL|Goodpasture antigen-binding protein|START domain containing 11|GPBP|Collagen type IV alpha-3-binding protein|Goodpasture antigen binding protein|StAR related lipid transfer (START) domain containing 11|StAR-related lipid transfer protein 11|Lipid transfer protein CERTL|START domain containing protein 11|Ceramide transfer protein|StARD11|Collagen type IV alpha 3 (Goodpasture antigen) binding protein|StAR related lipid transfer protein 11|START domain-containing protein 11
- Cert protein|FLJ20597|C43BP_HUMAN|Ceramide transporter|col4a3bp|hCERT|Alpha 3 type IV collagen binding protein|StARD 11|Procollagen type IV alpha 3 (Goodpasture antigen) binding protein|CERT|Collagen type IV alpha 3 binding protein|CERTL|Goodpasture antigen-binding protein|START domain containing 11|GPBP|Collagen type IV alpha-3-binding protein|Goodpasture antigen binding protein|StAR related lipid transfer (START) domain containing 11|StAR-related lipid transfer protein 11|Lipid transfer protein CERTL|START domain containing protein 11|Ceramide transfer protein|StARD11|Collagen type IV alpha 3 (Goodpasture antigen) binding protein|StAR related lipid transfer protein 11|START domain-containing protein 11
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5P4
- Q9Y5P4
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human COL4A3BP
- Fusion protein of Human COL4A3BP
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together