Order Entry
United States
ContactUsLinkComponent
Anti-NECAB3 Rabbit Polyclonal Antibody
Anti-NECAB3 Rabbit Polyclonal Antibody
Catalog # 77710-158
Supplier:  AFG Bioscience
New Product
Anti-NECAB3 Rabbit Polyclonal Antibody
Catalog # 77710-158
Supplier:  AFG Bioscience
Supplier Number:  A45845-100UL
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antigen Symbol:
    NECAB3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µl
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    Q96P71
  • Antigen Synonyms:
    NECAB3|Amyloid beta A4 protein-binding family A member 2-binding protein|Nek2-interacting protein 1|family A|SYTIP2|X11L-binding protein 51|dJ63M2.5|N-terminal EF-hand calcium-binding protein 3|N terminal EF hand calcium binding protein 3|EFCBP3|XB51|APBA2BP|member 2 binding protein|STIP3|Neuronal calcium binding protein 3|NECA3_HUMAN|Synaptotagmin interacting protein 2|X11L binding protein 51|Amyloid beta A4 protein binding family A member 2 binding protein|NIP1|amyloid beta (A4) precursor protein binding|Nek2 interacting protein 1|dJ63M2.4|Neuronal calcium-binding protein 3
  • Storage Buffer:
    PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
  • Storage Temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Synthesized peptide derived from internal of Human NECAB3
  • Purification:
    Antigen affinity purified
  • Cat. No.:
    77710-158
  • Packaging:
    Vial

Specifications

About this item

WB:1:500-1:3000, IF:1:100-1:500

Type:
Antigen: NECAB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse