Anti-PSMD11 Rabbit Polyclonal Antibody
Catalog # 77706-297
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:PSMD11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O00231
- Antigen Synonyms:26S proteasome regulatory subunit S9|26S proteasome regulatory subunit RPN6|PSD11_HUMAN|PSMD11|proteasome (prosome, macropain) 26S subunit, non-ATPase, 11|PSMD 11|Rpn6|MGC3844|proteasome 26S subunit, 11|26S proteasome regulatory subunit p44.5|protease 26S, subunit, 9|p44.5|26S proteasome non-ATPase regulatory subunit 11|S9|26S proteasome regulatory subunit 9
- Storage Buffer:PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human PSMD11
- Purification:Antigen affinity purified
- Cat. No.:77706-297
- Packaging:Vial
Specifications
About this item
WB:1:500-1:3000
Type:
Antigen: PSMD11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse