Aspiration Pipette
Catalog # 77589-552
Supplier: SARSTEDT INC
New Product
Specifications
- Capacity:2 ml
- Description:Aspiration pipette with tip, unplugged, sterile
- Environmentally Preferable:
- Cat. No.:77589-552
Specifications
About this item
Dosing, transferring, and measuring liquids are routine procedures in almost every laboratory. SARSTEDT offers a wide range of products for every application, from high-quality pipette tips and serological pipettes to transfer pipettes, for reliable and reproducible results.
- Working volume: 2 ml
- Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
- Polystyrene material
- Without graduations
The Aspiration Pipette is designed for transferring liquid with no need for precision, and for the aspiration of liquids using a vacuum pump. This pipette is designed without print and supplied without cotton plug.