Orion™ Sidekick Compressed Gas Ionizer, Simco Ion
Supplier: Simco
Certificates
About this item
Ionizer allows hands-free operation, which is ideal for assembly and manufacturing processes
The 120V model is UL and C-UL listed. The 230V model is CE marked.
A flexible mount allows the operator to focus the ionization airflow where it is needed. A foot pedal controls ionization. Airflow is adjustable. Balanced to ±15V. Operates on compressed air or nitrogen. Air input: 5–100psi.
Ordering information: Supplied with a hands-free stand and a steel bracket for bench mounting.
Frequently Bought Together