Aerostat™ XC Extended Coverage Ionizing Blower, Simco Ion
Supplier: Avantor
Certificates
About this item
Ionizing blower delivers broad coverage, stability, reliability, and rapid static discharge
The 120V model is UL and C-UL listed. The 230V model is CE marked.
No calibration is necessary; the blower’s inherent balance method maintains a balance of ±5V. A built-in emitter point cleaner provides easy maintenance. Emitter points never need replacing. A heater is provided for user comfort and a three-speed fan delivers a selectable, controlled airflow of 33L/s (70cfm), 42L/s (90cfm), or 57L/s (120cfm).
Frequently Bought Together