Order Entry
United States
ContactUsLinkComponent
Pierce™ Controlled Protein-Protein Cross-Linking Kit, Thermo Scientific
Pierce™ Controlled Protein-Protein Cross-Linking Kit, Thermo Scientific
Catalog # PI23456
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Controlled Protein-Protein Cross-Linking Kit, Thermo Scientific
Catalog # PI23456
Supplier:  Invitrogen
Supplier Number:  23456
CAS Number:  

Specifications

  • Host:
  • Description:
    Controlled Protein-Protein Cross-Linking Kit
  • Environmentally Preferable:
  • Cat. No.:
    PI23456
  • No. of reactions:
    1
  • Supplier no.:
    23456

Specifications

About this item

The Pierce™ Controlled Protein-Protein Crosslinking Kit contains essential crosslinkers, modification reagents and buffers for conjugating antibodies, enzymes and other purified proteins needed as probes.They insert an unbreakable molecular bridge between two proteins with easy-to-control stepwise coupling.

  • Reliable non cleavable heterobifunctional crosslinking agent offers proven chemistry, highly stable intermediates and efficient formation of the target conjugate
  • Includes disulfide (S-S) reductants and thiolation reagents
  • Ellman’s Reagent provides the option to monitor reaction sufficiently

The protocol is logically presented to make the procedure easy for both novices and experts. Chemicals in the kit were selected to allow for the most effective crosslinking. Reagents include a non-cleavable heterobifunctional crosslinking agent for stability and disulfide reductants and thiolation reagents to allow all possibilites of -SH groups to be coupled. A colorimetric reagent is also included to allow for optional reaction efficiency determination.

Delivery: The kit includes Sulfo-SMCC crosslinking agent (2 mg); 10X Activation/Conjugation Buffer (20 mL); BupH Phosphate Buffered Saline (2 packs); 2-Mercaptoethylamine-HCI (6 mg); Immobilised reductant (0.2 mL); SATA (2 mg); Dimethylformamide (DMF, 1 mL); Hydroxylamine-HCI (5 mg); Dextran Desalting Columns (2x10 mL); Column extender (2 each); Cysteine-HCI (20 mg); and Ellman’s reagent (2 mg).