To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Phospholipase A1
- Antigen Symbol:PLA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Pig,Human
- Western Blot:Yes
- Size:200 µl
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Gene ID:51365
- Antigen Synonyms:NMD|ps-PLA1|Phosphatidylserine-Specific Phospholipase A1-Alpha|PLA1A
- Amino Acid Number:309 - 452
- UniProtKB:Q53H76
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:19 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant PLA1 (Leu309~Lys452)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB151HU2
- Packaging:Vial
Polyclonal Antibody to Phospholipase A1 (PLA1), derived from recombinant PLA1 (Leu309~Lys452), is reactive with Human/Pig.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Phospholipase A1 is a phospholipase enzyme which removes the 1-acyl. Phospholipase A1 is a component of snake venom, but it also serves in normal metabolism to produce fatty acids from phospholipids. In melanocytic cells the Phospholipase A1 gene may be regulated by MITF. Phosphatidylserine-specific phospholipase A1-alpha (PLA1A) acts specifically on phosphatidylserine (PS) and 1-acyl-2-lysophosphatidylserine (lyso-PS) to hydrolyze fatty acids at the sn-1 position of these phospholipids. Although PLA1A belongs to the phospholipase gene family, the predicted amino acid sequence of the rat gene product demonstrated greater homology with mammalian triglyceride lipases than with phospholipases.
Type: Primary
Antigen: PLA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig