To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Kidney Injury Molecule 1
- Antigen Symbol:Kim1
- Clonality:Monoclonal
- Clone:C2
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:200 µl
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Gene ID:26762
- Antigen Synonyms:TIM1|HAVCR|T-cell immunoglobulin mucin receptor 1|HAVCR1|Hepatitis A Virus Cellular Receptor 1|TIMD1|T Cell Immunoglobulin And Mucin Domain-Containing Protein 1
- Amino Acid Number:21 - 240
- UniProtKB:Q96D42
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:54 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant Kim1 (Ser21~Ala240)
- Purification:Protein A + Protein G affinity chromatography
- Cat. No.:MSPP-MAA785HU2
- Packaging:Vial
Specifications
About this item
Monoclonal Antibody to Kidney Injury Molecule 1 (Kim1), derived from recombinant Kim1 (Ser21~Ala240), is reactive with Human.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Kidney injury molecule-1 (KIM-1) is a type 1 transmembrane protein that is not detectable in normal kidney tissue but is expressed at high levels in rodent kidneys with dedifferentiated proximal tubule epithelial cells after ischemic or toxic injury. Sequence analysis predicted that the 359-amino acid type I HAVCR1 glycoprotein, which is 79% identical to the monkey protein, contains a signal sequence; a 109-amino acid cys-rich N-terminal region with only 1 conserved N-glycosylation site; a 163-amino acid TSP-rich region with 3 N-glycosylation sites, two of which are conserved; and a 48-amino acid intracellular domain, 12 residues shorter than that of the monkey protein. Northern blot analysis revealed ubiquitous expression of a 4.4-kb HAVCR1 transcript, with highest levels in kidney and testis.
Type: Primary
Antigen: Kim1
Clonality: Monoclonal
Clone: C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human